Assay Details
| Gene Symbol: CG8176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104253.3 | NP_001097723.1 | 7 - 8 | 2404 | 57 | |
| NM_001104254.2 | NP_001097724.2 | 7 - 8 | 2175 | 57 | ||
| NM_001275490.1 | NP_001262419.1 | 7 - 8 | 1998 | 57 | ||
| NM_176435.3 | NP_788612.1 | 6 - 7 | 2100 | 57 | ||
| NM_176436.2 | NP_788613.2 | 6 - 7 | 1914 | 57 | ||
More Information
Set Membership: |
|

CG8176
Genomic Map