Assay Details
| Gene Symbol: CG8176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104253.3 | NP_001097723.1 | 10 - 11 | 3653 | 83 | |
| NM_001104254.2 | NP_001097724.2 | 10 - 11 | 3424 | 83 | ||
| NM_001275490.1 | NP_001262419.1 | 10 - 11 | 3247 | 83 | ||
| NM_176435.3 | NP_788612.1 | 9 - 10 | 3349 | 83 | ||
| NM_176436.2 | NP_788613.2 | 9 - 10 | 3163 | 83 | ||
| GenBank mRNA | BT015313.1 | - | 10 - 11 | 3317 | 83 | |
More Information
Set Membership: |
|

CG8176
Genomic Map