Assay Details
| Gene Symbol: Cad86C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260102.1 | NP_001247031.1 | 10 - 11 | 3265 | 66 | |
| NM_001260103.1 | NP_001247032.1 | 9 - 10 | 3181 | 66 | ||
| NM_001260104.1 | NP_001247033.1 | 10 - 11 | 3283 | 66 | ||
| NM_176458.4 | NP_788635.3 | 10 - 11 | 3921 | 66 | ||
More Information
Set Membership: |
|

Cad86C
Genomic Map