Assay Details
| Gene Symbol: B52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014619.3 | NP_001014619.2 | 7 - 8 | 943 | 78 | |
| NM_001170132.2 | NP_001163603.1 | 7 - 8 | 2926 | 78 | ||
| NM_001170133.2 | NP_001163604.1 | 6 - 7 | 3329 | 78 | ||
| NM_176488.3 | NP_788665.1 | 7 - 8 | 928 | 78 | ||
| NM_176489.3 | NP_788666.1 | 6 - 7 | 920 | 78 | ||
| NM_176490.3 | NP_788667.2 | 7 - 8 | 1088 | 78 | ||
| NM_176491.3 | NP_788668.1 | 7 - 8 | 943 | 78 | ||
| NM_176492.4 | NP_788669.1 | 8 - 9 | 2408 | 78 | ||
| NM_176493.4 | NP_788670.1 | 5 - 6 | 3321 | 78 | ||
| NM_176494.4 | NP_788671.2 | 7 - 8 | 953 | 78 | ||
| GenBank mRNA | AY113327.1 | - | 7 - 8 | 939 | 78 | |
| BT001495.1 | - | 7 - 8 | 856 | 78 | ||
| BT003286.1 | - | 7 - 8 | 879 | 78 | ||
| BT125783.1 | - | 7 - 8 | 940 | 78 | ||
More Information
Set Membership: |
|

B52
Genomic Map