Assay Details
| Gene Symbol: eIF4EHP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202346.2 | NP_001189275.1 | 2 - 3 | 951 | 57 | |
| NM_176552.3 | NP_788729.1 | 2 - 3 | 951 | 57 | ||
| GenBank mRNA | AY094966.1 | - | 2 - 3 | 935 | 57 | |
More Information
Set Membership: |
|

eIF4EHP
Genomic Map