Assay Details
| Gene Symbol: Antp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206445.1 | NP_996167.1 | 4 - 5 | 2387 | 82 | |
| NM_206446.1 | NP_996168.1 | 5 - 6 | 2176 | 82 | ||
| NM_206448.1 | NP_996170.1 | 4 - 5 | 2387 | 82 | ||
| NM_206449.1 | NP_996171.1 | 5 - 6 | 2176 | 82 | ||
| NM_206452.1 | NP_996174.1 | 4 - 5 | 2387 | 82 | ||
| NM_206453.1 | NP_996175.1 | 5 - 6 | 2176 | 82 | ||
| NM_206454.1 | NP_996176.1 | 4 - 5 | 2387 | 82 | ||
| GenBank mRNA | AY060407.1 | - | 4 - 5 | 2363 | 82 | |
| M20704.1 | - | 5 - 6 | 2182 | 82 | ||
| M20705.1 | - | 4 - 5 | 1907 | 82 | ||
More Information
Set Membership: |
|

Antp
Genomic Map