Assay Details
| Gene Symbol: Fer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275405.1 | NP_001262334.1 | 3 - 4 | 829 | 76 | |
| NM_206455.3 | NP_996177.1 | 3 - 4 | 814 | 76 | ||
| GenBank mRNA | AY089664.1 | - | 3 - 4 | 813 | 76 | |
| BT029276.1 | - | 784 | 76 | |||
More Information
Set Membership: |
|

Fer1
Genomic Map