Assay Details
| Gene Symbol: PK1-R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014620.2 | NP_001014620.1 | 1 - 2 | 980 | 59 | |
| NM_001260160.1 | NP_001247089.1 | 1 - 2 | 1265 | 59 | ||
| GenBank mRNA | AF368273.2 | - | 1 - 2 | 996 | 59 | |
| AF522191.1 | - | 1 - 2 | 860 | 59 | ||
More Information
Set Membership: |
|

PK1-R
Genomic Map