Assay Details
| Gene Symbol: Ada2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014634.3 | NP_001014634.1 | 1 - 2 | 172 | 65 | |
| NM_001014636.3 | NP_001014636.1 | 1 - 2 | 172 | 65 | ||
More Information
Set Membership: |
|

Ada2a
Genomic Map