Assay Details
| Gene Symbol: Stat92E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014643.2 | NP_001014643.1 | 5 - 6 | 2362 | 66 | |
| NM_001275828.1 | NP_001262757.1 | 5 - 6 | 2206 | 66 | ||
| NM_001275829.1 | NP_001262758.1 | 5 - 6 | 2362 | 66 | ||
| NM_001275831.1 | NP_001262760.1 | 6 - 7 | 2775 | 66 | ||
| NM_001275832.1 | NP_001262761.1 | 5 - 6 | 2206 | 66 | ||
| NM_001275833.1 | NP_001262762.1 | 6 - 7 | 2775 | 66 | ||
| NM_206521.2 | NP_996243.1 | 6 - 7 | 2619 | 66 | ||
| GenBank mRNA | BT021314.1 | - | 6 - 7 | 2620 | 66 | |
| U40070.1 | - | 6 - 7 | 2594 | 66 | ||
More Information
Set Membership: |
|

Stat92E
Genomic Map