Assay Details
| Gene Symbol: p53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170223.1 | NP_001163694.1 | 3 - 4 | 750 | 74 | |
| NM_206544.2 | NP_996267.1 | 3 - 4 | 525 | 74 | ||
| NM_206545.2 | NP_996268.1 | 2 - 3 | 232 | 74 | ||
| GenBank mRNA | AF192555.1 | - | 2 - 3 | 216 | 74 | |
| AF224713.1 | - | 2 - 3 | 209 | 74 | ||
| AF224714.1 | - | 2 - 3 | 216 | 74 | ||
| AF244918.1 | - | 2 - 3 | 216 | 74 | ||
| AF250918.1 | - | 2 - 3 | 216 | 74 | ||
| AF263722.1 | - | 2 - 3 | 232 | 74 | ||
| AY069096.1 | - | 2 - 3 | 216 | 74 | ||
| DQ191318.1 | - | 3 - 4 | 465 | 74 | ||
More Information
Set Membership: |
|

p53
Genomic Map