Assay Details
| Gene Symbol: p53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170223.1 | NP_001163694.1 | 6 - 7 | 1128 | 68 | |
| NM_001260323.2 | NP_001247252.2 | 2 - 3 | 357 | 68 | ||
| NM_206544.2 | NP_996267.1 | 6 - 7 | 903 | 68 | ||
| NM_206545.2 | NP_996268.1 | 5 - 6 | 610 | 68 | ||
| GenBank mRNA | AF192555.1 | - | 5 - 6 | 594 | 68 | |
| AF224713.1 | - | 5 - 6 | 587 | 68 | ||
| AF224714.1 | - | 5 - 6 | 594 | 68 | ||
| AF244918.1 | - | 5 - 6 | 594 | 68 | ||
| AF250918.1 | - | 5 - 6 | 594 | 68 | ||
| AF263722.1 | - | 5 - 6 | 610 | 68 | ||
| AY069096.1 | - | 5 - 6 | 594 | 68 | ||
| DQ191318.1 | - | 6 - 7 | 843 | 68 | ||
More Information
Set Membership: |
|

p53
Genomic Map