Assay Details
| Gene Symbol: slo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014651.3 | NP_001014651.1 | 12 - 13 | 2420 | 89 | |
| NM_001014652.3 | NP_001014652.1 | 12 - 13 | 2420 | 89 | ||
| NM_001014653.3 | NP_001014653.1 | 12 - 13 | 2420 | 89 | ||
| NM_001014660.3 | NP_001014660.1 | 12 - 13 | 2420 | 89 | ||
| NM_001275995.1 | NP_001262924.1 | 14 - 15 | 2350 | 89 | ||
| NM_001275997.1 | NP_001262926.1 | 13 - 14 | 2161 | 89 | ||
| NM_001275998.1 | NP_001262927.1 | 13 - 14 | 2161 | 89 | ||
| NM_001275999.1 | NP_001262928.1 | 13 - 14 | 2161 | 89 | ||
| NM_206566.4 | NP_996289.1 | 13 - 14 | 2161 | 89 | ||
More Information
Set Membership: |
|

slo
Genomic Map