Assay Details
| Gene Symbol: Pxd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170162.2 | NP_001163633.1 | 7 - 8 | 1927 | 81 | |
| NM_001275725.1 | NP_001262654.1 | 7 - 8 | 2248 | 81 | ||
| NM_206501.3 | NP_996223.1 | 7 - 8 | 2254 | 81 | ||
More Information
Set Membership: |
|

Pxd
Genomic Map