Assay Details
| Gene Symbol: Bsg25D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273163.2 | NP_001260092.1 | 1 - 2 | 591 | 136 | |
| NM_057568.6 | NP_476916.4 | 1 - 2 | 591 | 136 | ||
More Information
Set Membership: |
|

Bsg25D
Genomic Map