Assay Details
| Gene Symbol: Pdi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300141.1 | NP_001287070.1 | 1 - 2 | 537 | 125 | |
| NM_079355.3 | NP_524079.1 | 1 - 2 | 289 | 125 | ||
| GenBank mRNA | BT001544.1 | - | 1 - 2 | 277 | 125 | |
| U18973.1 | - | 1 - 2 | 159 | 125 | ||
More Information
Set Membership: |
|

Pdi
Genomic Map