Assay Details
| Gene Symbol: Obp56c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166374.4 | NP_725925.3 | 1 - 2 | 129 | 143 | |
| GenBank mRNA | BT100151.4 | - | 1 - 2 | 73 | 143 | |
More Information
Set Membership: |
|

Obp56c
Genomic Map