Assay Details
| Gene Symbol: dpy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258946.1 | NP_001245875.1 | 11 - 12 | 11313 | 130 | |
| NM_001258947.3 | NP_001245876.1 | 13 - 14 | 11531 | 130 | ||
| NM_001258950.1 | NP_001245879.1 | 9 - 10 | 3678 | 130 | ||
| NM_001273101.1 | NP_001260030.1 | 12 - 13 | 11303 | 130 | ||
| NM_001273102.1 | NP_001260031.1 | 11 - 12 | 11313 | 130 | ||
| NM_001273103.1 | NP_001260032.1 | 11 - 12 | 11313 | 130 | ||
| NM_001273104.1 | NP_001260033.1 | 10 - 11 | 10956 | 130 | ||
| NM_001273105.1 | NP_001260034.1 | 11 - 12 | 11313 | 130 | ||
| NM_001273106.1 | NP_001260035.1 | 11 - 12 | 11313 | 130 | ||
| NM_001273107.1 | NP_001260036.1 | 11 - 12 | 11313 | 130 | ||
| NM_001273108.1 | NP_001260037.1 | 9 - 10 | 3678 | 130 | ||
| NM_001273109.1 | NP_001260038.1 | 12 - 13 | 11303 | 130 | ||
| NM_001273110.1 | NP_001260039.1 | 12 - 13 | 11303 | 130 | ||
| NM_001273113.1 | NP_001260042.1 | 11 - 12 | 11313 | 130 | ||
More Information
Set Membership: |
|

dpy
Genomic Map