Assay Details
| Gene Symbol: dpy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258946.1 | NP_001245875.1 | 15 - 16 | 23630 | 148 | |
| NM_001258947.3 | NP_001245876.1 | 17 - 18 | 23848 | 148 | ||
| NM_001273101.1 | NP_001260030.1 | 16 - 17 | 23620 | 148 | ||
| NM_001273102.1 | NP_001260031.1 | 15 - 16 | 23630 | 148 | ||
| NM_001273103.1 | NP_001260032.1 | 15 - 16 | 23630 | 148 | ||
| NM_001273104.1 | NP_001260033.1 | 14 - 15 | 23273 | 148 | ||
| NM_001273106.1 | NP_001260035.1 | 15 - 16 | 23630 | 148 | ||
| NM_001273107.1 | NP_001260036.1 | 15 - 16 | 23630 | 148 | ||
| NM_001273108.1 | NP_001260037.1 | 13 - 14 | 15995 | 148 | ||
| NM_001273109.1 | NP_001260038.1 | 16 - 17 | 23620 | 148 | ||
| NM_001273110.1 | NP_001260039.1 | 16 - 17 | 23620 | 148 | ||
| NM_001273113.1 | NP_001260042.1 | 15 - 16 | 23630 | 148 | ||
More Information
Set Membership: |
|

dpy
Genomic Map