Assay Details
| Gene Symbol: dpy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258946.1 | NP_001245875.1 | 29 - 30 | 31441 | 116 | |
| NM_001258947.3 | NP_001245876.1 | 31 - 32 | 31659 | 116 | ||
| NM_001258950.1 | NP_001245879.1 | 22 - 23 | 10264 | 116 | ||
| NM_001273101.1 | NP_001260030.1 | 29 - 30 | 30813 | 116 | ||
| NM_001273102.1 | NP_001260031.1 | 29 - 30 | 31441 | 116 | ||
| NM_001273103.1 | NP_001260032.1 | 29 - 30 | 31441 | 116 | ||
| NM_001273104.1 | NP_001260033.1 | 28 - 29 | 31084 | 116 | ||
| NM_001273105.1 | NP_001260034.1 | 25 - 26 | 18517 | 116 | ||
| NM_001273108.1 | NP_001260037.1 | 27 - 28 | 23806 | 116 | ||
| NM_001273109.1 | NP_001260038.1 | 28 - 29 | 30591 | 116 | ||
| NM_001273110.1 | NP_001260039.1 | 28 - 29 | 30591 | 116 | ||
| NM_001273111.1 | NP_001260040.1 | 23 - 24 | 16869 | 116 | ||
| NM_001273112.1 | NP_001260041.1 | 22 - 23 | 16879 | 116 | ||
More Information
Set Membership: |
|

dpy
Genomic Map