Assay Details
| Gene Symbol: dpy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258946.1 | NP_001245875.1 | 31 - 32 | 32090 | 139 | |
| NM_001258947.3 | NP_001245876.1 | 33 - 34 | 32308 | 139 | ||
| NM_001258950.1 | NP_001245879.1 | 24 - 25 | 10913 | 139 | ||
| NM_001273101.1 | NP_001260030.1 | 31 - 32 | 31462 | 139 | ||
| NM_001273102.1 | NP_001260031.1 | 31 - 32 | 32090 | 139 | ||
| NM_001273103.1 | NP_001260032.1 | 31 - 32 | 32090 | 139 | ||
| NM_001273104.1 | NP_001260033.1 | 30 - 31 | 31733 | 139 | ||
| NM_001273105.1 | NP_001260034.1 | 27 - 28 | 19166 | 139 | ||
| NM_001273108.1 | NP_001260037.1 | 29 - 30 | 24455 | 139 | ||
| NM_001273109.1 | NP_001260038.1 | 30 - 31 | 31240 | 139 | ||
| NM_001273110.1 | NP_001260039.1 | 30 - 31 | 31240 | 139 | ||
| NM_001273111.1 | NP_001260040.1 | 25 - 26 | 17518 | 139 | ||
| NM_001273112.1 | NP_001260041.1 | 24 - 25 | 17528 | 139 | ||
More Information
Set Membership: |
|

dpy
Genomic Map