Assay Details
| Gene Symbol: dpy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273101.1 | NP_001260030.1 | 42 - 43 | 39461 | 148 | |
| NM_001273103.1 | NP_001260032.1 | 42 - 43 | 40089 | 148 | ||
| NM_001273104.1 | NP_001260033.1 | 41 - 42 | 39732 | 148 | ||
| NM_001273105.1 | NP_001260034.1 | 38 - 39 | 27165 | 148 | ||
| NM_001273106.1 | NP_001260035.1 | 36 - 37 | 38142 | 148 | ||
| NM_001273107.1 | NP_001260036.1 | 31 - 32 | 36213 | 148 | ||
| NM_001273108.1 | NP_001260037.1 | 40 - 41 | 32454 | 148 | ||
| NM_001273109.1 | NP_001260038.1 | 41 - 42 | 39239 | 148 | ||
| NM_001273111.1 | NP_001260040.1 | 36 - 37 | 25517 | 148 | ||
| NM_001273112.1 | NP_001260041.1 | 35 - 36 | 25527 | 148 | ||
| NM_001273113.1 | NP_001260042.1 | 31 - 32 | 36213 | 148 | ||
More Information
Set Membership: |
|

dpy
Genomic Map