Assay Details
| Gene Symbol: dpy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273101.1 | NP_001260030.1 | 44 - 45 | 40413 | 147 | |
| NM_001273103.1 | NP_001260032.1 | 44 - 45 | 41041 | 147 | ||
| NM_001273104.1 | NP_001260033.1 | 43 - 44 | 40684 | 147 | ||
| NM_001273105.1 | NP_001260034.1 | 40 - 41 | 28117 | 147 | ||
| NM_001273106.1 | NP_001260035.1 | 38 - 39 | 39094 | 147 | ||
| NM_001273107.1 | NP_001260036.1 | 33 - 34 | 37165 | 147 | ||
| NM_001273108.1 | NP_001260037.1 | 42 - 43 | 33406 | 147 | ||
| NM_001273109.1 | NP_001260038.1 | 43 - 44 | 40191 | 147 | ||
| NM_001273111.1 | NP_001260040.1 | 38 - 39 | 26469 | 147 | ||
| NM_001273112.1 | NP_001260041.1 | 37 - 38 | 26479 | 147 | ||
| NM_001273113.1 | NP_001260042.1 | 33 - 34 | 37165 | 147 | ||
More Information
Set Membership: |
|

dpy
Genomic Map