Assay Details
| Gene Symbol: dpy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273101.1 | NP_001260030.1 | 72 - 73 | 59452 | 86 | |
| NM_001273102.1 | NP_001260031.1 | 56 - 57 | 53363 | 86 | ||
| NM_001273103.1 | NP_001260032.1 | 72 - 73 | 60080 | 86 | ||
| NM_001273104.1 | NP_001260033.1 | 71 - 72 | 59723 | 86 | ||
| NM_001273105.1 | NP_001260034.1 | 68 - 69 | 47156 | 86 | ||
| NM_001273106.1 | NP_001260035.1 | 66 - 67 | 58133 | 86 | ||
| NM_001273107.1 | NP_001260036.1 | 61 - 62 | 56204 | 86 | ||
| NM_001273108.1 | NP_001260037.1 | 70 - 71 | 52445 | 86 | ||
| NM_001273111.1 | NP_001260040.1 | 66 - 67 | 45508 | 86 | ||
More Information
Set Membership: |
|

dpy
Genomic Map