Assay Details
| Gene Symbol: dpy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258946.1 | NP_001245875.1 | 7 - 8 | 1824 | 104 | |
| NM_001258947.3 | NP_001245876.1 | 9 - 10 | 2042 | 104 | ||
| NM_001273101.1 | NP_001260030.1 | 8 - 9 | 1814 | 104 | ||
| NM_001273102.1 | NP_001260031.1 | 7 - 8 | 1824 | 104 | ||
| NM_001273103.1 | NP_001260032.1 | 7 - 8 | 1824 | 104 | ||
| NM_001273105.1 | NP_001260034.1 | 7 - 8 | 1824 | 104 | ||
| NM_001273106.1 | NP_001260035.1 | 7 - 8 | 1824 | 104 | ||
| NM_001273107.1 | NP_001260036.1 | 7 - 8 | 1824 | 104 | ||
| NM_001273109.1 | NP_001260038.1 | 8 - 9 | 1814 | 104 | ||
| NM_001273110.1 | NP_001260039.1 | 8 - 9 | 1814 | 104 | ||
| NM_001273111.1 | NP_001260040.1 | 8 - 9 | 1814 | 104 | ||
| NM_001273112.1 | NP_001260041.1 | 7 - 8 | 1824 | 104 | ||
| NM_001273113.1 | NP_001260042.1 | 7 - 8 | 1824 | 104 | ||
More Information
Set Membership: |
|

dpy
Genomic Map