Assay Details
| Gene Symbol: Ctr1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143580.3 | NP_651837.1 | 1 | 626 | 225 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctr1C| Gene Symbol: Ctr1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143580.3 | NP_651837.1 | 1 | 626 | 225 | |
Set Membership: |
|