Assay Details
| Gene Symbol: Mal-A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165608.2 | NP_724679.2 | 1 - 2 | 1155 | 141 | |
| NM_206057.3 | NP_995779.2 | 1 - 2 | 1155 | 141 | ||
More Information
Set Membership: |
|

Mal-A6
Genomic Map