Assay Details
| Gene Symbol: Antp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382007.1 | NP_001368996.1 | 3 - 4 | 1469 | 144 | |
| NM_206444.1 | NP_996166.1 | 3 - 4 | 2354 | 144 | ||
| NM_206447.1 | NP_996169.1 | 3 - 4 | 2354 | 144 | ||
| NM_206450.1 | NP_996172.1 | 4 - 5 | 2143 | 144 | ||
| NM_206451.1 | NP_996173.1 | 4 - 5 | 2143 | 144 | ||
More Information
Set Membership: |
|

Antp
Genomic Map