Assay Details
| Gene Symbol: Antp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206444.1 | NP_996166.1 | 4 - 5 | 2560 | 117 | |
| NM_206449.1 | NP_996171.1 | 6 - 7 | 2388 | 117 | ||
| NM_206451.1 | NP_996173.1 | 5 - 6 | 2349 | 117 | ||
| NM_206452.1 | NP_996174.1 | 5 - 6 | 2599 | 117 | ||
More Information
Set Membership: |
|

Antp
Genomic Map