Assay Details
| Gene Symbol: twe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273562.2 | NP_001260491.1 | 2 | 1697 | 134 | |
| NM_001273563.1 | NP_001260492.1 | 2 | 1671 | 134 | ||
| NM_001273564.2 | NP_001260493.1 | 2 | 1810 | 134 | ||
| NM_057285.4 | NP_476633.1 | 2 | 1810 | 134 | ||
| GenBank mRNA | AY061266.1 | - | 2 | 1800 | 134 | |
| M94158.1 | - | 2 | 1628 | 134 | ||
More Information
Set Membership: |
|

twe
Genomic Map