Assay Details
| Gene Symbol: pn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057336.5 | NP_476684.1 | 2 | 1429 | 152 | |
| GenBank mRNA | AY051502.1 | - | 2 | 1537 | 152 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pn| Gene Symbol: pn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057336.5 | NP_476684.1 | 2 | 1429 | 152 | |
| GenBank mRNA | AY051502.1 | - | 2 | 1537 | 152 | |
Set Membership: |
|