Assay Details
| Gene Symbol: eEF1alpha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299392.1 | NP_001286321.1 | 1 | 1735 | 61 | |
| NM_001299393.1 | NP_001286322.1 | 2 | 1832 | 61 | ||
| NM_058027.5 | NP_477375.1 | 2 | 1785 | 61 | ||
| NM_165850.3 | NP_725085.1 | 2 | 1792 | 61 | ||
| GenBank mRNA | BT011039.1 | - | 2458 | 61 | ||
| BT099573.1 | - | 2 | 1783 | 61 | ||
| Gene Symbol: PHGPx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BT053727.1 | - | 270 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BT053727.1 transcripts.
Set Membership: |
|

eEF1alpha1
Genomic Map