Assay Details
| Gene Symbol: eEF1alpha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165850.3 | NP_725085.1 | 2 | 2091 | 153 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

eEF1alpha1| Gene Symbol: eEF1alpha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165850.3 | NP_725085.1 | 2 | 2091 | 153 | |
Set Membership: |
|