Assay Details
| Gene Symbol: SkpA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038729.3 | NP_001033818.1 | 1 | 1371 | 127 | |
| NM_001297826.1 | NP_001284755.1 | 1 | 653 | 127 | ||
| NM_058042.5 | NP_477390.1 | 2 | 914 | 127 | ||
| NM_166856.3 | NP_726690.1 | 2 | 762 | 127 | ||
| NM_166857.3 | NP_726691.1 | 2 | 877 | 127 | ||
| NM_166858.3 | NP_726692.1 | 2 | 1299 | 127 | ||
| NM_166859.3 | NP_726693.1 | 2 | 696 | 127 | ||
| NM_166860.3 | NP_726694.1 | 2 | 740 | 127 | ||
| NM_166861.3 | NP_726695.1 | 2 | 672 | 127 | ||
| GenBank mRNA | AY069297.1 | - | 1 | 1333 | 127 | |
| BT099582.1 | - | 2 | 700 | 127 | ||
| BT099610.1 | - | 2 | 857 | 127 | ||
More Information
Set Membership: |
|

SkpA
Genomic Map