Assay Details
| Gene Symbol: Taf10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058070.6 | NP_477418.1 | 1 | 464 | 98 | |
| GenBank mRNA | AJ237968.1 | - | 1 | 405 | 98 | |
| AJ243837.1 | - | 2 | 666 | 98 | ||
| AY071220.1 | - | 1 | 440 | 98 | ||
More Information
Set Membership: |
|

Taf10b
Genomic Map