Assay Details
| Gene Symbol: fs(1)Ya | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058145.4 | NP_477493.2 | 2 | 2135 | 88 | |
| GenBank mRNA | AY069542.1 | - | 2 | 2117 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fs(1)Ya| Gene Symbol: fs(1)Ya | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058145.4 | NP_477493.2 | 2 | 2135 | 88 | |
| GenBank mRNA | AY069542.1 | - | 2 | 2117 | 88 | |
Set Membership: |
|