Assay Details
| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274987.1 | NP_001261916.1 | 2 | 1482 | 64 | |
| NM_001274988.1 | NP_001261917.1 | 3 | 2032 | 64 | ||
| NM_001274989.1 | NP_001261918.1 | 2 | 1961 | 64 | ||
| NM_079377.3 | NP_524101.2 | 2 | 1746 | 64 | ||
| NM_168644.2 | NP_730097.1 | 2 | 1598 | 64 | ||
| NM_168645.2 | NP_730098.1 | 2 | 1548 | 64 | ||
| GenBank mRNA | BT031197.1 | - | 2 | 1580 | 64 | |
More Information
Set Membership: |
|

Diap1
Genomic Map