Assay Details
| Gene Symbol: Mst84Dc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104231.2 | NP_001097701.1 | 2 | 292 | 74 | |
| NM_079530.4 | NP_524254.1 | 2 | 296 | 74 | ||
| GenBank mRNA | BT023340.1 | - | 2 | 269 | 74 | |
More Information
Set Membership: |
|

Mst84Dc
Genomic Map