Assay Details
| Gene Symbol: sra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300432.1 | NP_001287361.1 | 2 | 1905 | 107 | |
| NM_079654.3 | NP_524378.1 | 2 | 1689 | 107 | ||
| GenBank mRNA | AF147700.1 | - | 2 | 1709 | 107 | |
| AY061194.1 | - | 2 | 1710 | 107 | ||
More Information
Set Membership: |
|

sra
Genomic Map