Assay Details
| Gene Symbol: CG10939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079973.3 | NP_524712.1 | 2 | 1539 | 127 | |
| GenBank mRNA | AY047533.1 | - | 2 | 1521 | 127 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG10939| Gene Symbol: CG10939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079973.3 | NP_524712.1 | 2 | 1539 | 127 | |
| GenBank mRNA | AY047533.1 | - | 2 | 1521 | 127 | |
Set Membership: |
|