Assay Details
| Gene Symbol: tara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080255.3 | NP_524994.2 | 2 | 3368 | 61 | |
| NM_169709.2 | NP_732120.1 | 2 | 3093 | 61 | ||
| GenBank mRNA | BT021316.1 | - | 2 | 3002 | 61 | |
| BT099632.1 | - | 2 | 3089 | 61 | ||
More Information
Set Membership: |
|

tara
Genomic Map