Assay Details
| Gene Symbol: rib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274155.1 | NP_001261084.1 | 2 | 4186 | 116 | |
| NM_001274156.2 | NP_001261085.1 | 2 | 3898 | 116 | ||
| NM_080534.4 | NP_536795.1 | 2 | 3898 | 116 | ||
| GenBank mRNA | AF416603.1 | - | 2 | 3881 | 116 | |
| BT010295.1 | - | 2 | 3881 | 116 | ||
More Information
Set Membership: |
|

rib
Genomic Map