Assay Details
| Gene Symbol: Rtca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297872.1 | NP_001284801.1 | 2 | 1929 | 131 | |
| NM_130621.3 | NP_569977.1 | 2 | 1957 | 131 | ||
| GenBank mRNA | AY052112.1 | - | 2 | 1957 | 131 | |
More Information
Set Membership: |
|

Rtca
Genomic Map