Assay Details
| Gene Symbol: Gbeta13F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316638.2 | NP_001303567.1 | 2 | 1817 | 61 | |
| NM_080351.7 | NP_525090.1 | 3 | 1670 | 61 | ||
| NM_206736.2 | NP_996459.1 | 3 | 1591 | 61 | ||
| NM_206737.2 | NP_996460.1 | 4 | 1832 | 61 | ||
| NM_206738.3 | NP_996461.1 | 3 | 1276 | 61 | ||
| NM_206739.2 | NP_996462.1 | 3 | 1507 | 61 | ||
| GenBank mRNA | AY058566.1 | - | 3 | 1613 | 61 | |
More Information
Set Membership: |
|

Gbeta13F
Genomic Map