Assay Details
| Gene Symbol: spi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032110.2 | NP_001027281.1 | 3 | 1180 | 110 | |
| NM_057561.6 | NP_476909.2 | 2 | 1061 | 110 | ||
| NM_134291.5 | NP_599118.2 | 3 | 1257 | 110 | ||
| NM_134292.3 | NP_599119.2 | 2 | 1042 | 110 | ||
| NM_134293.3 | NP_599120.2 | 3 | 1238 | 110 | ||
| GenBank mRNA | BT044503.1 | - | 2 | 1071 | 110 | |
More Information
Set Membership: |
|

spi
Genomic Map