Assay Details
| Gene Symbol: brat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201913.2 | NP_001188842.2 | 4 | 3291 | 64 | |
| NM_001299154.1 | NP_001286083.1 | 3 | 2901 | 64 | ||
| NM_057597.4 | NP_476945.1 | 4 | 3291 | 64 | ||
| NM_134302.2 | NP_599129.1 | 3 | 2995 | 64 | ||
| NM_206004.2 | NP_995726.1 | 3 | 2929 | 64 | ||
| GenBank mRNA | AB022432.1 | - | 4 | 3256 | 64 | |
| AF119332.1 | - | 3 | 2960 | 64 | ||
| BT125989.1 | - | 4 | 3302 | 64 | ||
More Information
Set Membership: |
|

brat
Genomic Map