Assay Details
| Gene Symbol: Dhod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057876.5 | NP_477224.1 | 2 | 1173 | 53 | |
| NM_134311.3 | NP_599138.3 | 2 | 1290 | 53 | ||
| GenBank mRNA | BT009968.1 | - | 2 | 1164 | 53 | |
More Information
Set Membership: |
|

Dhod
Genomic Map