Assay Details
| Gene Symbol: meng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298759.1 | NP_001285688.1 | 2 | 2035 | 152 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

meng| Gene Symbol: meng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298759.1 | NP_001285688.1 | 2 | 2035 | 152 | |
Set Membership: |
|