Assay Details
| Gene Symbol: Nfs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135689.2 | NP_609533.1 | 2 | 1475 | 87 | |
| GenBank mRNA | AY113363.1 | - | 2 | 1475 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nfs1| Gene Symbol: Nfs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135689.2 | NP_609533.1 | 2 | 1475 | 87 | |
| GenBank mRNA | AY113363.1 | - | 2 | 1475 | 87 | |
Set Membership: |
|