Assay Details
| Gene Symbol: CG13085 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273645.1 | NP_001260574.1 | 2 | 837 | 105 | |
| NM_136120.3 | NP_609964.1 | 2 | 1075 | 105 | ||
More Information
Set Membership: |
|

CG13085
Genomic Map